Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212501.2 | NP_997666.1 | 10 - 11 | 1130 | 67 | |
XM_006255920.2 | XP_006255982.1 | 10 - 11 | 1179 | 67 | ||
GenBank mRNA | BC083703.1 | - | 8 - 9 | 1356 | 67 | |
BC127469.1 | - | 10 - 11 | 1102 | 7324889 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |