Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009178.2 | NP_001009178.1 | 3 - 4 | 711 | 59 | |
GenBank mRNA | BC158566.1 | - | 3 - 4 | 711 | 9092936 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |