Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100895.1 | NP_001094365.1 | 4 - 5 | 588 | 106 | |
GenBank mRNA | BC091222.1 | - | 4 - 5 | 562 | 106 | |
BC105853.1 | - | 4 - 5 | 585 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |