Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108117.1 | NP_001101587.1 | 4 - 5 | 314 | 65 | |
GenBank mRNA | FQ226451.1 | - | 4 - 5 | 290 | 65 | |
FQ234671.1 | - | 4 - 5 | 290 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |