Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008324.1 | NP_001008325.1 | 5 - 6 | 552 | 72 | |
GenBank mRNA | BC085933.1 | - | 5 - 6 | 552 | 7112408 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |