Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034941.1 | NP_001030113.1 | 6 - 7 | 713 | 58 | |
XM_006242428.2 | XP_006242490.1 | 6 - 7 | 793 | 58 | ||
XM_006242429.2 | XP_006242491.1 | 5 - 6 | 606 | 58 | ||
XM_006242430.2 | XP_006242492.1 | 4 - 5 | 451 | 58 | ||
GenBank mRNA | BC104690.1 | - | 6 - 7 | 713 | 7107060 | 58 |
FQ228813.1 | - | 6 - 7 | 707 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |