Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030998.1 | NP_112260.1 | 5 - 6 | 691 | 82 | |
GenBank mRNA | KJ923229.1 | - | 5 - 6 | 691 | 82 | |
U42427.1 | - | 5 - 6 | 691 | 82 | ||
X71916.1 | - | 5 - 6 | 673 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |