Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106797.3 | NP_001100267.1 | 4 - 5 | 513 | 132 | |
XM_006242391.2 | XP_006242453.1 | 4 - 5 | 577 | 132 | ||
XM_006242393.2 | XP_006242455.1 | 3 - 4 | 472 | 132 | ||
XM_008765707.1 | XP_008763929.1 | 3 - 4 | 486 | 132 | ||
GenBank mRNA | BC166908.1 | - | 4 - 5 | 514 | 6889335 | 132 |
BC168739.1 | - | 4 - 5 | 514 | 6918617 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |