Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106797.3 | NP_001100267.1 | 2 - 3 | 142 | 157 | |
XM_006242391.2 | XP_006242453.1 | 2 - 3 | 206 | 157 | ||
GenBank mRNA | BC166908.1 | - | 2 - 3 | 143 | 6889335 | 157 |
BC168739.1 | - | 2 - 3 | 143 | 6918617 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |