Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013106.1 | NP_001013124.1 | 2 - 3 | 196 | 71 | |
GenBank mRNA | BC091366.1 | - | 2 - 3 | 196 | 7319183 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |