Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106794.2 | NP_001100264.2 | 4 - 5 | 328 | 118 | |
GenBank mRNA | BC169128.1 | - | 4 - 5 | 294 | 6921287 | 118 |
CF976389.1 | - | 4 - 5 | 332 | 118 | ||
CF976393.1 | - | 4 - 5 | 299 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |