Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024388.2 | NP_077364.2 | 3 - 4 | 1123 | 85 | |
XM_006242356.2 | XP_006242418.1 | 3 - 4 | 3950 | 85 | ||
XM_006242358.2 | XP_006242420.1 | 3 - 4 | 1878 | 85 | ||
GenBank mRNA | BC097313.1 | - | 3 - 4 | 1116 | 7458540 | 85 |
U17254.1 | - | 3 - 4 | 1116 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |