Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271369.1 | NP_001258298.1 | 10 - 11 | 3772 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |