Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191710.1 | NP_001178639.1 | 3 - 4 | 1855 | 141 | |
XM_006233082.2 | XP_006233144.1 | 3 - 4 | 1554 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |