Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025119.1 | NP_001020290.1 | 2 - 3 | 310 | 97 | |
GenBank mRNA | BC091420.1 | - | 2 - 3 | 310 | 7365370 | 97 |
FQ230444.1 | - | 1 - 2 | 119 | 97 | ||
FQ230757.1 | - | 1 - 2 | 160 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |