Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107694.1 | NP_001101164.1 | 6 - 7 | 473 | 88 | |
XM_006232876.2 | XP_006232938.1 | 5 - 6 | 1971 | 88 | ||
XM_006232878.2 | XP_006232940.1 | 5 - 6 | 2280 | 88 | ||
XM_008761300.1 | XP_008759522.1 | 4 - 5 | 722 | 88 | ||
XM_008761301.1 | XP_008759523.1 | 5 - 6 | 2226 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |