Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107694.1 | NP_001101164.1 | 9 - 10 | 880 | 64 | |
XM_006232876.2 | XP_006232938.1 | 8 - 9 | 2378 | 64 | ||
XM_006232878.2 | XP_006232940.1 | 8 - 9 | 2687 | 64 | ||
XM_008761300.1 | XP_008759522.1 | 7 - 8 | 1129 | 64 | ||
XM_008761301.1 | XP_008759523.1 | 8 - 9 | 2633 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |