Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rorc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232926.2 | XP_006232988.2 | 5 - 6 | 860 | 116 | |
XM_008761339.1 | XP_008759561.1 | 4 - 5 | 3184 | 116 |
Gene Symbol: Lingo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109189.1 | NP_001102659.1 | 5 - 6 | 1023 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Nuclear Receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |