Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025118.1 | NP_001020289.1 | 3 - 4 | 770 | 59 | |
XM_006232886.2 | XP_006232948.1 | 6 - 7 | 1994 | 59 | ||
XM_008761304.1 | XP_008759526.1 | 7 - 8 | 1952 | 59 | ||
GenBank mRNA | BC091386.1 | - | 3 - 4 | 770 | 7321358 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |