Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108559.1 | NP_001102029.1 | 6 - 7 | 885 | 101 | |
XM_006232968.2 | XP_006233030.1 | 6 - 7 | 893 | 101 | ||
XM_006232969.2 | XP_006233031.1 | 6 - 7 | 2567 | 101 | ||
XM_006232970.1 | XP_006233032.1 | 6 - 7 | 859 | 101 | ||
XM_008761313.1 | XP_008759535.1 | 6 - 7 | 894 | 101 | ||
XM_008761314.1 | XP_008759536.1 | 6 - 7 | 895 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |