Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001072193.5 | XP_001072193.3 | 10 - 11 | 1973 | 102 | |
XM_006232937.2 | XP_006232999.1 | 10 - 11 | 1973 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |