Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107709.1 | NP_001101179.1 | 1 - 2 | 512 | 96 | |
XM_006233081.2 | XP_006233143.1 | 2 - 3 | 379 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |