Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107709.1 | NP_001101179.1 | 8 - 9 | 1902 | 62 | |
XM_006233081.2 | XP_006233143.1 | 9 - 10 | 1769 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |