Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011951.1 | NP_001011951.1 | 1 - 2 | 269 | 76 | |
GenBank mRNA | BC078997.1 | - | 1 - 2 | 269 | 7112738 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |