Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025684.1 | NP_001020855.1 | 3 - 4 | 542 | 93 | |
GenBank mRNA | BC098865.1 | - | 3 - 4 | 542 | 7380579 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |