Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173094.2 | NP_775117.2 | 2 - 3 | 588 | 76 | |
XM_006233002.2 | XP_006233064.1 | 2 - 3 | 617 | 76 | ||
GenBank mRNA | BC078695.1 | - | 2 - 3 | 588 | 7112644 | 76 |
BC083543.1 | - | 2 - 3 | 588 | 7132009 | 76 | |
M33648.1 | - | 2 - 3 | 606 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |