Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080766.2 | NP_542944.1 | 6 - 7 | 725 | 65 | |
GenBank mRNA | BC098659.1 | - | 6 - 7 | 725 | 7368145 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |