Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106452.1 | NP_001099922.1 | 9 - 10 | 1646 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |