Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdadc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012156.1 | NP_001012156.1 | 1 - 2 | 166 | 140 | |
XM_006252123.1 | XP_006252185.1 | 1 - 2 | 214 | 140 | ||
XM_006252125.2 | XP_006252187.1 | 1 - 2 | 214 | 140 | ||
XM_008770760.1 | XP_008768982.1 | 1 - 2 | 216 | 140 | ||
GenBank mRNA | BC083736.1 | - | 1 - 2 | 166 | 7191631 | 140 |
FQ213068.1 | - | 1 - 2 | 185 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |