Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011917.1 | NP_001011917.1 | 2 - 3 | 206 | 142 | |
XM_006252098.2 | XP_006252160.1 | 3 - 4 | 310 | 142 | ||
XM_008770742.1 | XP_008768964.1 | 4 - 5 | 403 | 142 | ||
GenBank mRNA | BC083684.1 | - | 2 - 3 | 206 | 7188949 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |