Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011917.1 | NP_001011917.1 | 3 - 4 | 343 | 70 | |
XM_006252098.2 | XP_006252160.1 | 4 - 5 | 447 | 70 | ||
XM_006252099.2 | XP_006252161.1 | 5 - 6 | 531 | 70 | ||
XM_006252100.2 | XP_006252162.1 | 6 - 7 | 727 | 70 | ||
XM_006252101.2 | XP_006252163.1 | 5 - 6 | 643 | 70 | ||
XM_008770742.1 | XP_008768964.1 | 5 - 6 | 540 | 70 | ||
XM_008770743.1 | XP_008768965.1 | 2 - 3 | 937 | 70 | ||
GenBank mRNA | BC083684.1 | - | 3 - 4 | 343 | 7188949 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |