Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083337.1 | NP_001076806.1 | 2 - 3 | 135 | 153 | |
XM_006252181.2 | XP_006252243.1 | 2 - 3 | 1038 | 153 | ||
GenBank mRNA | BC158553.1 | - | 2 - 3 | 234 | 9088212 | 153 |
EF446165.1 | - | 2 - 3 | 135 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |