Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270855.1 | NP_001257784.1 | 5 - 6 | 583 | 96 | |
NM_001270856.1 | NP_001257785.1 | 4 - 5 | 502 | 96 | ||
NM_001270857.1 | NP_001257786.1 | 4 - 5 | 499 | 96 | ||
NM_001270858.1 | NP_001257787.1 | 4 - 5 | 502 | 96 | ||
NM_001270859.1 | NP_001257788.1 | 5 - 6 | 514 | 96 | ||
NM_019176.2 | NP_062049.1 | 5 - 6 | 583 | 96 | ||
XM_006252149.2 | XP_006252211.1 | 6 - 7 | 737 | 96 | ||
XM_006252151.2 | XP_006252213.1 | 5 - 6 | 1646 | 96 | ||
XM_006252156.2 | XP_006252218.1 | 4 - 5 | 1558 | 96 | ||
XM_008770778.1 | XP_008769000.1 | 5 - 6 | 587 | 96 | ||
GenBank mRNA | AF026528.1 | - | 4 - 5 | 443 | 96 | |
AF026529.1 | - | 4 - 5 | 443 | 96 | ||
AF026530.1 | - | 5 - 6 | 524 | 96 | ||
BC092646.1 | - | 5 - 6 | 520 | 7319130 | 96 | |
CB580507.1 | - | 4 - 5 | 433 | 96 | ||
FQ212720.1 | - | 3 - 4 | 313 | 96 | ||
FQ212976.1 | - | 4 - 5 | 453 | 96 | ||
FQ213134.1 | - | 4 - 5 | 503 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |