Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139091.2 | NP_620791.1 | 9 - 10 | 1072 | 100 | |
XM_006252093.2 | XP_006252155.1 | 9 - 10 | 1177 | 100 | ||
GenBank mRNA | AF000900.1 | - | 8 - 9 | 943 | 100 | |
BC085690.1 | - | 9 - 10 | 1072 | 7190650 | 100 | |
U63839.1 | - | 9 - 10 | 1085 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |