Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053999.2 | NP_446451.1 | 2 - 3 | 379 | 92 | |
XM_008770730.1 | XP_008768952.1 | 2 - 3 | 1186 | 92 | ||
GenBank mRNA | BC128731.1 | - | 2 - 3 | 379 | 7370343 | 92 |
M83297.1 | - | 2 - 3 | 274 | 92 | ||
M83298.1 | - | 2 - 3 | 360 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |