Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpine3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252195.2 | XP_006252257.1 | 2 - 3 | 527 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |