Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037181.1 | NP_001032258.1 | 3 - 4 | 446 | 132 | |
XM_006253087.2 | XP_006253149.1 | 2 - 3 | 275 | 132 | ||
XM_006253089.2 | XP_006253151.1 | 3 - 4 | 510 | 132 | ||
XM_008771223.1 | XP_008769445.1 | 3 - 4 | 705 | 132 | ||
GenBank mRNA | BC107470.1 | - | 3 - 4 | 446 | 7457548 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |