Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022926.1 | NP_075215.1 | 7 - 8 | 1310 | 124 | |
XM_006253083.1 | XP_006253145.1 | 7 - 8 | 1335 | 124 | ||
GenBank mRNA | AF076167.1 | - | 7 - 8 | 1310 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |