Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039027.1 | NP_001034116.1 | 8 - 9 | 1109 | 121 | |
XM_006240906.1 | XP_006240968.1 | 8 - 9 | 1186 | 121 | ||
GenBank mRNA | BC099828.1 | - | 8 - 9 | 1109 | 7934398 | 121 |
FQ212635.1 | - | 8 - 9 | 1112 | 121 | ||
FQ212816.1 | - | 8 - 9 | 1112 | 121 | ||
FQ219525.1 | - | 8 - 9 | 1113 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |