Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039849.2 | NP_001034938.1 | 1 - 2 | 267 | 100 | |
GenBank mRNA | AB072798.1 | - | 1 - 2 | 267 | 100 | |
AF274028.1 | - | 1 - 2 | 276 | 100 | ||
AY570513.1 | - | 1 - 2 | 255 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |