Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138509.3 | NP_612518.2 | 5 - 6 | 677 | 84 | |
XM_006235294.2 | XP_006235356.1 | 5 - 6 | 660 | 84 | ||
XM_006235295.2 | XP_006235357.1 | 5 - 6 | 730 | 84 | ||
GenBank mRNA | BC081726.1 | - | 5 - 6 | 677 | 7192469 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |