Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101005.1 | NP_001094475.1 | 3 - 4 | 482 | 59 | |
XM_006235253.2 | XP_006235315.1 | 2 - 3 | 475 | 59 | ||
GenBank mRNA | BC083575.1 | - | 3 - 4 | 482 | 7112877 | 59 |
FQ209572.1 | - | 2 - 3 | 435 | 59 | ||
FQ225908.1 | - | 2 - 3 | 372 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |