Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107795.1 | NP_001101265.1 | 3 - 4 | 439 | 75 | |
XM_006235404.2 | XP_006235466.1 | 4 - 5 | 515 | 75 | ||
XM_008762342.1 | XP_008760564.1 | 4 - 5 | 526 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |