Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107795.1 | NP_001101265.1 | 8 - 9 | 1294 | 118 | |
XM_006235404.2 | XP_006235466.1 | 9 - 10 | 1370 | 118 | ||
XM_008762342.1 | XP_008760564.1 | 9 - 10 | 1381 | 118 | ||
GenBank mRNA | FQ226444.1 | - | 2 - 3 | 255 | 118 | |
FQ231010.1 | - | 2 - 3 | 255 | 118 | ||
FQ232478.1 | - | 2 - 3 | 255 | 118 | ||
FQ232700.1 | - | 4 - 5 | 878 | 118 | ||
FQ232914.1 | - | 5 - 6 | 826 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |