Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005887.1 | NP_001005887.1 | 10 - 11 | 1182 | 147 | |
XM_008762335.1 | XP_008760557.1 | 12 - 13 | 1410 | 147 | ||
XM_008762336.1 | XP_008760558.1 | 12 - 13 | 1335 | 147 | ||
XM_008762337.1 | XP_008760559.1 | 9 - 10 | 1087 | 147 | ||
GenBank mRNA | AY600518.1 | - | 10 - 11 | 1182 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |