Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005887.1 | NP_001005887.1 | 18 - 19 | 2073 | 86 | |
XM_008762335.1 | XP_008760557.1 | 20 - 21 | 2301 | 86 | ||
XM_008762336.1 | XP_008760558.1 | 20 - 21 | 2226 | 86 | ||
XM_008762337.1 | XP_008760559.1 | 17 - 18 | 1978 | 86 | ||
GenBank mRNA | AY600518.1 | - | 18 - 19 | 2073 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |