Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsfl1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031981.2 | NP_114187.1 | 4 - 5 | 552 | 78 | |
XM_006235254.2 | XP_006235316.1 | 5 - 6 | 525 | 78 | ||
GenBank mRNA | AB002086.1 | - | 4 - 5 | 552 | 78 | |
BC072464.1 | - | 4 - 5 | 449 | 7096463 | 78 | |
FQ228953.1 | - | 5 - 6 | 490 | 78 | ||
Y10769.1 | - | 4 - 5 | 449 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |