Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107790.1 | NP_001101260.1 | 10 - 11 | 1584 | 112 | |
XM_006235270.2 | XP_006235332.1 | 11 - 12 | 1487 | 112 | ||
XM_006235272.2 | XP_006235334.1 | 11 - 12 | 1439 | 112 | ||
XM_006235273.2 | XP_006235335.1 | 10 - 11 | 1369 | 112 | ||
XM_006235274.2 | XP_006235336.1 | 11 - 12 | 1487 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |