Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014051.1 | NP_001014073.1 | 5 - 6 | 482 | 68 | |
XM_006235276.2 | XP_006235338.1 | 5 - 6 | 1253 | 68 | ||
XM_008762331.1 | XP_008760553.1 | 6 - 7 | 537 | 68 | ||
XM_008762332.1 | XP_008760554.1 | 6 - 7 | 537 | 68 | ||
GenBank mRNA | BC082105.1 | - | 5 - 6 | 482 | 7134735 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |