Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfa2t2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168542.1 | NP_001162014.1 | 6 - 7 | 1064 | 97 | |
XM_006235311.2 | XP_006235373.1 | 6 - 7 | 1048 | 97 | ||
XM_006235312.2 | XP_006235374.1 | 7 - 8 | 1076 | 97 | ||
XM_008762313.1 | XP_008760535.1 | 6 - 7 | 1152 | 97 | ||
GenBank mRNA | FQ222452.1 | - | 6 - 7 | 1080 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |